3-Ethyl-3-hydroxyazetidine hydrochloride

Supplier: Matrix Scientific

Synonyms: 3-Ethylazetidin-3-ol hydrochloride

113029-1G 113029-5G
102891-396EA 930.77 USD
102891-396 102891-398
3-Ethyl-3-hydroxyazetidine hydrochloride
3-Ethyl-3-hydroxyazetidine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO
MW: 137.61 g/mol
MDL Number: MFCD11849053
CAS Number: 935668-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR