3,3-Dimethylazetidine hydrochloride

Supplier: Matrix Scientific

113028-1G 113028-5G
102891-392EA 727.69 USD
102891-392 102891-394
3,3-Dimethylazetidine hydrochloride
3,3-Dimethylazetidine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClN
MDL Number: MFCD10565785
CAS Number: 89381-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR