1-Diphenylmethyl-3-methyleneazetidine

Supplier: Matrix Scientific

113024-1G 113024-5G
102891-382EA 256.92 USD
102891-382 102891-384
1-Diphenylmethyl-3-methyleneazetidine
1-Diphenylmethyl-3-methyleneazetidine

Hazard: Irritant

Formula: C₁₇H₁₇N
MDL Number: MFCD11559000
CAS Number: 40569-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR