1-Benzyl-3,3-difluoro-4,4-piperidinediol

Supplier: Matrix Scientific

Synonyms: 1-Benzyl-3,3-difluoropiperidine-4,4-diol

113022-1G 113022-5G
102891-374EA 727.69 USD
102891-374 102891-376
1-Benzyl-3,3-difluoro-4,4-piperidinediol
1-Benzyl-3,3-difluoro-4,4-piperidinediol

Hazard: Irritant

Formula: C₁₂H₁₅F₂NO₂
MW: 243.26 g/mol
MDL Number: MFCD20926176
CAS Number: 1067914-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR