2-Azabicyclo[2.2.2]octan-3-one

Supplier: Matrix Scientific

113017-1G 113017-5G
102897-574EA 727.69 USD
102897-574 102897-576
2-Azabicyclo[2.2.2]octan-3-one
2-Azabicyclo[2.2.2]octan-3-one

Hazard: Irritant

Formula: C₇H₁₁NO
MDL Number: MFCD16547516
CAS Number: 3306-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR