1-Oxa-7-azaspiro[3.5]nonane oxalate
Supplier: Matrix Scientific
112987-1G
112987-5G
102891-324EA
378.46
USD
102891-324
102891-326
1-Oxa-7-azaspiro[3.5]nonane oxalate
1-Oxa-7-azaspiro[3.5]nonane oxalate
Hazard: Irritant
Formula:
C₉H₁₅NO₅ |
MDL Number:
MFCD22575226 CAS Number: 1408076-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...