1-Boc-3-methylpyrrolidine-3-methanol

Supplier: Matrix Scientific

112978-1G 112978-5G
102891-306EA 727.69 USD
102891-306 102891-308
1-Boc-3-methylpyrrolidine-3-methanol
1-Boc-3-methylpyrrolidine-3-methanol

Hazard: Irritant

Formula: C₁₁H₂₁NO₃
MDL Number: MFCD20926173
CAS Number: 1263506-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR