1-Boc-3-methylpyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 1-(tert-Butoxycarbonyl)-3-methylpyrrolidine-3-carboxylic acid

112977-1G 112977-5G
102891-302EA 640 USD
102891-302 102891-304
1-Boc-3-methylpyrrolidine-3-carboxylic acid
1-Boc-3-methylpyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD15071411
CAS Number: 887587-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR