Methyl-1-boc-3-methylpyrrolidine-3-carboxylate

Supplier: Matrix Scientific

112976-1G 112976-5G
102891-298EA 640 USD
102891-298 102891-300
Methyl-1-boc-3-methylpyrrolidine-3-carboxylate
Methyl-1-boc-3-methylpyrrolidine-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₁NO₄
MDL Number: MFCD22415188
CAS Number: 457657-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR