3,5-Dibromoaniline ≥98%
Supplier: Matrix Scientific
087315-5G
087315-25G
102853-296EA
60
USD
102853-296
102853-298
3,5-Dibromoaniline ≥98%
3,5-Dibromoaniline
Hazard: Irritant-Harmful/Refrigerate/Light Sensitive
Formula:
C₆H₅Br₂N Boiling Pt: 296 °C Melting Pt: 55…58 °C Density: 2.022g/cc Flash Pt: 133 °C |
MDL Number:
MFCD00047841 CAS Number: 626-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...