(Triisopropylsilyl)acetylene ≥97%

Supplier: Matrix Scientific

Synonyms: Ethynyltriisopropylsilane

087204-5G 087204-25G
102852-900EA 38.46 USD
102852-900 102852-902
(Triisopropylsilyl)acetylene ≥97%
(Triisopropylsilyl)acetylene

Hazard: Flammable/Refrigerate

Formula: C₁₁H₂₂Si
MW: 182.38 g/mol
Boiling Pt: 50…53 °C (0.6 mmHg)
Density: 0.813
Flash Pt: 56 °C
MDL Number: MFCD00075452
CAS Number: 89343-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR