tert-Butyl-4-(methoxymethyl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

087302-500MG 087302-1G
102853-280EA 803.08 USD
102853-280 102853-282
tert-Butyl-4-(methoxymethyl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(methoxymethyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₃NO₃
MW: 229.32 g/mol
MDL Number: MFCD07779470
CAS Number: 509147-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR