tert-Butyl-4-(3-bromo-5-nitropyridin-4-yl)piperazine-1-carboxylate ≥95%
Supplier: Matrix Scientific
087298-500MG
087298-1G
102853-268EA
298.46
USD
102853-268
102853-270
tert-Butyl-4-(3-bromo-5-nitropyridin-4-yl)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(3-bromo-5-nitropyridin-4-yl)piperazine-1-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₉BrN₄O₄ MW: 387.24 g/mol Melting Pt: 106…107 °C |
MDL Number:
MFCD22573528 CAS Number: 1407516-48-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...