tert-Butyl-5-benzyl-octahydropyrrolo[3,4-c]pyrrole-2-carboxylate ≥97%

Supplier: Matrix Scientific

087297-500MG 087297-1G
102853-264EA 803.08 USD
102853-264 102853-266
tert-Butyl-5-benzyl-octahydropyrrolo[3,4-c]pyrrole-2-carboxylate ≥97%
tert-Butyl-5-benzyl-octahydropyrrolo[3,4-c]pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₆N₂O₂
MW: 302.42 g/mol
MDL Number: MFCD18909636
CAS Number: 186202-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR