tert-Butyl-11-azatricyclo[6.2.1.0

Supplier: Matrix Scientific

087296-500MG 087296-1G
102853-260EA 803.08 USD
102853-260 102853-262
tert-Butyl-11-azatricyclo[6.2.1.0
tert-Butyl-11-azatricyclo[6.2.1.0^{2,7}]undeca- 2,4,6,9-tetraene-11-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₇NO₂
MW: 243.31 g/mol
MDL Number: MFCD22573547
CAS Number: 5176-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR