Estrone acetate ≥97%

Supplier: Matrix Scientific

087294-500MG 087294-1G
102853-250EA 193.85 USD
102853-250 102853-252
Estrone acetate ≥97%
Estrone acetate

Hazard: Irritant

Formula: C₂₀H₂₄O₃
MDL Number: MFCD00056411
CAS Number: 901-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR