4-Propanoylphenyl-2,2-dimethylpropanoate ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Propanoylphenyl 2,2-dimethylpropanoate

087292-500MG 087292-1G
102853-240EA 193.85 USD
102853-240 102853-242
4-Propanoylphenyl-2,2-dimethylpropanoate ≥97%
4-Propanoylphenyl-2,2-dimethylpropanoate

Hazard: Irritant

Formula: C₁₄H₁₈O₃
MW: 234.3 g/mol
MDL Number: MFCD01530702
CAS Number: 120703-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR