Methyl-7-(trifluoromethyl)-1-benzofuran-2-carboxylate ≥95%

Supplier: Matrix Scientific

087291-500MG 087291-1G
102853-236EA 304.62 USD
102853-236 102853-238
Methyl-7-(trifluoromethyl)-1-benzofuran-2-carboxylate ≥95%
Methyl-7-(trifluoromethyl)-1-benzofuran-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₇F₃O₃
MW: 244.17 g/mol
Melting Pt: 51…53 °C
MDL Number: MFCD22573541
CAS Number: 1407521-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR