Methyl-5-(trifluoromethyl)-1-benzofuran-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-(trifluoromethyl)benzofuran-2-carboxylate

087290-500MG 087290-1G
102853-232EA 600 USD
102853-232 102853-234
Methyl-5-(trifluoromethyl)-1-benzofuran-2-carboxylate ≥95%
Methyl-5-(trifluoromethyl)-1-benzofuran-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₇F₃O₃
MW: 244.17 g/mol
Melting Pt: 97…100 °C
MDL Number: MFCD22573538
CAS Number: 1407521-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR