6-Methyl-1,2,4-triazin-5-ol ≥97%

Supplier: Matrix Scientific

087289-500MG 087289-1G
102853-228EA 803.08 USD
102853-228 102853-230
6-Methyl-1,2,4-triazin-5-ol ≥97%
6-Methyl-1,2,4-triazin-5-ol

Hazard: Irritant

Formula: C₄H₅N₃O
MW: 111.1 g/mol
MDL Number: MFCD11108305
CAS Number: 16120-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR