Methyl-6-nitro-2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

087286-500MG 087286-1G
102853-218EA 803.08 USD
102853-218 102853-220
Methyl-6-nitro-2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carboxylate ≥95%
Methyl-6-nitro-2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carboxylate

Hazard: Irritant

Formula: C₉H₇N₃O₅
Melting Pt: >300 °C
MDL Number: MFCD22573536
CAS Number: 205259-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR