Methyl-2-(2-nitrobenzylidene)acetoacetate ≥95%

Supplier: Matrix Scientific

087285-500MG 087285-1G 087285-5G
102853-212EA 158.46 USD
102853-212 102853-214 102853-216
Methyl-2-(2-nitrobenzylidene)acetoacetate ≥95%
Methyl-2-(2-nitrobenzylidene)acetoacetate

Hazard: Irritant

Formula: C₁₂H₁₁NO₅
MW: 249.23 g/mol
MDL Number: MFCD00551860
CAS Number: 39562-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR