Methyl (2E)-2-(N-hydroxyimino)-3-(4-hydroxyphenyl)propanoate ≥97%

Supplier: Matrix Scientific

087283-500MG 087283-1G
102853-204EA 403.08 USD
102853-204 102853-206
Methyl (2E)-2-(N-hydroxyimino)-3-(4-hydroxyphenyl)propanoate ≥97%
Methyl (2E)-2-(N-hydroxyimino)-3-(4-hydroxyphenyl)propanoate

Hazard: Irritant

Formula: C₁₀H₁₁NO₄
Melting Pt: 131…132 °C
MDL Number: MFCD22573515
CAS Number: 50563-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR