Methyl-7-ethoxy-1-benzofuran-2-carboxylate ≥97%

Supplier: Matrix Scientific

087281-500MG 087281-1G
102853-196EA 193.85 USD
102853-196 102853-198
Methyl-7-ethoxy-1-benzofuran-2-carboxylate ≥97%
Methyl-7-ethoxy-1-benzofuran-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₂O₄
Melting Pt: 99…101 °C
MDL Number: MFCD22573539
CAS Number: 1407521-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR