Methyl-5-cyano-2-hydroxy-3-iodobenzoate ≥95%

Supplier: Matrix Scientific

087280-500MG 087280-1G
102853-192EA 600 USD
102853-192 102853-194
Methyl-5-cyano-2-hydroxy-3-iodobenzoate ≥95%
Methyl-5-cyano-2-hydroxy-3-iodobenzoate

Hazard: Irritant

Formula: C₉H₆INO₃
MW: 303.06 g/mol
Melting Pt: 184…185 °C
MDL Number: MFCD09951959
CAS Number: 952302-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR