Methyl-6-chloro-1-benzofuran-2-carboxylate ≥97%

Supplier: Matrix Scientific

087278-500MG 087278-1G
102853-184EA 600 USD
102853-184 102853-186
Methyl-6-chloro-1-benzofuran-2-carboxylate ≥97%
Methyl-6-chloro-1-benzofuran-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₇ClO₃
MW: 210.62 g/mol
Melting Pt: 97…99 °C
MDL Number: MFCD22123551
CAS Number: 323580-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR