Methyl (3E)-4-(2-bromophenyl)-2-oxobut-3-enoate ≥95%
Supplier: Matrix Scientific
087276-500MG
087276-1G
102853-178EA
600
USD
102853-178
102853-180
Methyl (3E)-4-(2-bromophenyl)-2-oxobut-3-enoate ≥95%
Methyl (3E)-4-(2-bromophenyl)-2-oxobut-3-enoate
Hazard: Irritant
Formula:
C₁₁H₉BrO₃ |
MDL Number:
MFCD22573543 CAS Number: 956476-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...