Methyl-5-bromo-6-(methylamino)pyridine-3-carboxylate ≥97%

Supplier: Matrix Scientific

087275-500MG 087275-1G
102853-174EA 600 USD
102853-174 102853-176
Methyl-5-bromo-6-(methylamino)pyridine-3-carboxylate ≥97%
Methyl-5-bromo-6-(methylamino)pyridine-3-carboxylate

Hazard: Irritant

Formula: C₈H₉BrN₂O₂
MW: 245.08 g/mol
Melting Pt: 125…127 °C
MDL Number: MFCD22371032
CAS Number: 1244016-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR