Methyl-7-bromo-1-benzofuran-2-carboxylate ≥90%

Supplier: Matrix Scientific

087271-500MG 087271-1G
102853-158EA 529.23 USD
102853-158 102853-160
Methyl-7-bromo-1-benzofuran-2-carboxylate ≥90%
Methyl-7-bromo-1-benzofuran-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₇BrO₃
MW: 255.07 g/mol
Melting Pt: 73…76 °C
MDL Number: MFCD14648669
CAS Number: 1247406-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR