Methyl-1-benzofuran-2-carboxylate ≥95%
Supplier: Matrix Scientific
087270-500MG
087270-1G
102853-154EA
193.85
USD
102853-154
102853-156
Methyl-1-benzofuran-2-carboxylate ≥95%
Methyl-1-benzofuran-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₈O₃ Melting Pt: 53…55 °C |
MDL Number:
MFCD01159618 CAS Number: 1646-27-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...