Methyl-6-amino-2,5-dichloropyrimidine-4-carboxylate ≥97%

Supplier: Matrix Scientific

087269-500MG 087269-1G
102853-150EA 604.62 USD
102853-150 102853-152
Methyl-6-amino-2,5-dichloropyrimidine-4-carboxylate ≥97%
Methyl-6-amino-2,5-dichloropyrimidine-4-carboxylate

Hazard: Irritant

Formula: C₆H₅Cl₂N₃O₂
MW: 222.03 g/mol
Melting Pt: 168…170 °C
MDL Number: MFCD16660543
CAS Number: 502142-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR