Methyl-5-acetyl-4-iodo-1H-pyrrole-2-carboxylate ≥97%

Supplier: Matrix Scientific

087268-500MG 087268-1G
102853-146EA 600 USD
102853-146 102853-148
Methyl-5-acetyl-4-iodo-1H-pyrrole-2-carboxylate ≥97%
Methyl-5-acetyl-4-iodo-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₈H₈INO₃
MW: 293.06 g/mol
Melting Pt: 121…122 °C
MDL Number: MFCD22573513
CAS Number: 1407516-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR