3-Methoxy-2-methylbenzohydrazide ≥97%

Supplier: Matrix Scientific

087267-500MG 087267-1G
102853-142EA 403.08 USD
102853-142 102853-144
3-Methoxy-2-methylbenzohydrazide ≥97%
3-Methoxy-2-methylbenzohydrazide

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MW: 180.21 g/mol
Melting Pt: 114…117 °C
MDL Number: MFCD12031987
CAS Number: 740799-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR