3-Iodoquinolin-8-amine ≥97%

Supplier: Matrix Scientific

087266-500MG 087266-1G
102853-138EA 600 USD
102853-138 102853-140
3-Iodoquinolin-8-amine ≥97%
3-Iodoquinolin-8-amine

Hazard: Irritant

Formula: C₉H₇IN₂
Melting Pt: 121…125 °C
MDL Number: MFCD09907947
CAS Number: 497084-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR