5-Iodo-2-methoxypyridine-3-carboxylic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
5-Iodo-2-methoxynicotinic acid
087264-500MG
087264-1G
102853-130EA
469.23
USD
102853-130
102853-132
5-Iodo-2-methoxypyridine-3-carboxylic acid ≥97%
5-Iodo-2-methoxypyridine-3-carboxylic acid
Hazard: Irritant
Formula:
C₇H₆INO₃ Melting Pt: 194…196 °C |
MDL Number:
MFCD22573540 CAS Number: 1407521-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...