1-(5-Iodo-6-methoxy-2-methylpyridin-3-yl)ethan-1-one ≥97%
Supplier: Matrix Scientific
087263-500MG
087263-1G
102853-126EA
995.38
USD
102853-126
102853-128
1-(5-Iodo-6-methoxy-2-methylpyridin-3-yl)ethan-1-one ≥97%
1-(5-Iodo-6-methoxy-2-methylpyridin-3-yl)ethan-1-one
Hazard: Irritant
Formula:
C₉H₁₀INO₂ Melting Pt: 111…112 °C |
MDL Number:
MFCD22573533 CAS Number: 1407516-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...