4-(4-Hydroxy-3-iodophenyl)benzonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 4'-Hydroxy-3'-iodo-[1,1'-biphenyl]-4-carbonitrile

087260-500MG 087260-1G
102853-112EA 600 USD
102853-112 102853-114
4-(4-Hydroxy-3-iodophenyl)benzonitrile ≥97%
4-(4-Hydroxy-3-iodophenyl)benzonitrile

Hazard: Irritant

Formula: C₁₃H₈INO
MW: 321.12 g/mol
MDL Number: MFCD06796628
CAS Number: 460746-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR