[1-(2-Fluorophenyl)-1H-pyrrol-2-yl]methanol ≥97%

Supplier: Matrix Scientific

087259-500MG 087259-1G
102853-108EA 448.5 USD
102853-108 102853-110
[1-(2-Fluorophenyl)-1H-pyrrol-2-yl]methanol ≥97%
[1-(2-Fluorophenyl)-1H-pyrrol-2-yl]methanol

Hazard: Irritant

Formula: C₁₁H₁₀FNO
Melting Pt: 37…40 °C
MDL Number: MFCD09754615
CAS Number: 132407-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR