1-[(4-Fluorophenyl)methyl]-1H-indazole-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

087257-500MG 087257-1G
102853-100EA 600 USD
102853-100 102853-102
1-[(4-Fluorophenyl)methyl]-1H-indazole-3-carboxylic acid ≥97%
1-[(4-Fluorophenyl)methyl]-1H-indazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₁FN₂O₂
MDL Number: MFCD01719717
CAS Number: 50264-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR