(4-Fluoro-3-iodophenyl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Fluoro-3-iodobenzyl alcohol

087253-500MG 087253-1G
102853-084EA 803.08 USD
102853-084 102853-086
(4-Fluoro-3-iodophenyl)methanol ≥95%
(4-Fluoro-3-iodophenyl)methanol

Hazard: Irritant

Formula: C₇H₆FIO
Melting Pt: 43…45 °C
MDL Number: MFCD18396949
CAS Number: 227609-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR