Ethyl-5-[(4-methylpiperazin-1-yl)methyl]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

087252-500MG 087252-1G
102853-080EA 600 USD
102853-080 102853-082
Ethyl-5-[(4-methylpiperazin-1-yl)methyl]pyridine-2-carboxylate ≥95%
Ethyl-5-[(4-methylpiperazin-1-yl)methyl]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₁N₃O₂
MW: 263.34 g/mol
Melting Pt: 62…64 °C
MDL Number: MFCD22573525
CAS Number: 1407532-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR