Ethyl-3-formylpyridine-2-carboxylate ≥97%

Supplier: Matrix Scientific

087250-500MG 087250-1G
102853-072EA 995.38 USD
102853-072 102853-074
Ethyl-3-formylpyridine-2-carboxylate ≥97%
Ethyl-3-formylpyridine-2-carboxylate

Hazard: Irritant

Formula: C₉H₉NO₃
MW: 179.18 g/mol
Melting Pt: 81…83 °C
MDL Number: MFCD14560545
CAS Number: 159755-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR