Ethyl-2,6-dichloropyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2,6-Dichloropyridine-3-carboxylate, 2,6-Dichloropyridine-3-carboxylic acid ethyl ester, 2,6-Dichloronicotinic acid ethyl ester, Ethyl 2,6-Dichloronicotinate

087249-500MG 087249-1G 087249-5G
102853-066EA 115.38 USD
102853-066 102853-068 102853-070
Ethyl-2,6-dichloropyridine-3-carboxylate ≥95%
Ethyl-2,6-dichloropyridine-3-carboxylate

Hazard: Irritant

Formula: C₈H₇Cl₂NO₂
MW: 220.06 g/mol
Melting Pt: 50…51 °C
MDL Number: MFCD09757510
CAS Number: 58584-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR