Ethyl-4-chloro-5-(trifluoromethyl)-pyridine-2-carboxylate ≥95%
Supplier: Matrix Scientific
087247-500MG
087247-1G
102853-058EA
1058.46
USD
102853-058
102853-060
Ethyl-4-chloro-5-(trifluoromethyl)-pyridine-2-carboxylate ≥95%
Ethyl-4-chloro-5-(trifluoromethyl)-pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₉H₇ClF₃NO₂ MW: 253.61 g/mol Melting Pt: 47…49 °C |
MDL Number:
MFCD22573527 CAS Number: 1407516-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...