1-(5-Ethyl-1-benzofuran-2-yl)ethan-1-one ≥97%

Supplier: Matrix Scientific

087246-500MG 087246-1G
102853-054EA 803.08 USD
102853-054 102853-056
1-(5-Ethyl-1-benzofuran-2-yl)ethan-1-one ≥97%
1-(5-Ethyl-1-benzofuran-2-yl)ethan-1-one

Hazard: Irritant

Formula: C₁₂H₁₂O₂
MDL Number: MFCD11574557
CAS Number: 106989-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR