(2,4-Dioxo-1,2,3,4-tetrahydropyrimidin-5-ylamino)fumaric acid dimethyl ester ≥97%
Supplier: Matrix Scientific
087245-500MG
087245-1G
102853-050EA
193.85
USD
102853-050
102853-052
(2,4-Dioxo-1,2,3,4-tetrahydropyrimidin-5-ylamino)fumaric acid dimethyl ester ≥97%
(2,4-Dioxo-1,2,3,4-tetrahydropyrimidin-5-ylamino)fumaric acid dimethyl ester
Hazard: Irritant
Formula:
C₁₀H₁₁N₃O₆ MW: 269.22 g/mol |
MDL Number:
MFCD22580727 CAS Number: 60458-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...