2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7-ol ≥95%

Supplier: Matrix Scientific

087244-500MG 087244-1G
102853-046EA 403.08 USD
102853-046 102853-048
2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7-ol ≥95%
2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7-ol

Hazard: Irritant

Formula: C₈H₉N₃O
Melting Pt: 251…253 °C
MDL Number: MFCD08729601
CAS Number: 27166-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR