2,4-Dimethyl-3-amino-1H-pyrrole-2,4-dicarboxylate ≥95%

Supplier: Matrix Scientific

087243-500MG 087243-1G 087243-5G
102853-040EA 403.08 USD
102853-040 102853-042 102853-044
2,4-Dimethyl-3-amino-1H-pyrrole-2,4-dicarboxylate ≥95%
2,4-Dimethyl-3-amino-1H-pyrrole-2,4-dicarboxylate

Hazard: Irritant

Formula: C₈H₁₀N₂O₄
MW: 198.18 g/mol
Melting Pt: 165…166 °C
MDL Number: MFCD10566520
CAS Number: 180059-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR