(1S)-1-(3,4-Difluorophenyl)propan-1-amine hydrochloride ≥97%

Supplier: Matrix Scientific

087242-500MG 087242-1G
102853-036EA 803.08 USD
102853-036 102853-038
(1S)-1-(3,4-Difluorophenyl)propan-1-amine hydrochloride ≥97%
(1S)-1-(3,4-Difluorophenyl)propan-1-amine hydrochloride

Hazard: Irritant

Formula: C₉H₁₂ClF₂N
MDL Number: MFCD12757716
CAS Number: 847448-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR