1,4-Diethyl-2-bromobenzene-1,4-dicarboxylate ≥97%

Supplier: Matrix Scientific

087241-500MG 087241-1G 087241-5G
102853-030EA 172.31 USD
102853-030 102853-032 102853-034
1,4-Diethyl-2-bromobenzene-1,4-dicarboxylate ≥97%
1,4-Diethyl-2-bromobenzene-1,4-dicarboxylate

Hazard: Irritant

Formula: C₁₂H₁₃BrO₄
MW: 301.14 g/mol
MDL Number: MFCD12913608
CAS Number: 154239-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR